USP HPLC Organic Impurity Analysis for Lamotrigine Tablets (SST) on an Ascentis® Express C18
![USP HPLC Organic Impurity Analysis for Lamotrigine Tablets (SST) on an Ascentis® Express C18](/deepweb/assets/sigmaaldrich/product/images/118/010/19ef433b-2dab-419d-9982-7572bdf2baf0/800/19ef433b-2dab-419d-9982-7572bdf2baf0.jpg)
Materials
analytical column
mobile phase component
sample prep
CONDITIONS
column
Ascentis® Express C18, 250 x 4.6 mm I.D., 5 µm (50538-U)
mobile phase
Acetonitrile:methanol:buffer (10:30:60) v/v/v; Buffer– 0.8 g/L of ammonium acetate, adjusted with glacial acetic acid to a pH of 4.5
gradient
Isocratic
flow rate
1 mL/min
pressure
216 bar (3132 psi)
column temp.
Ambient
detector
UV, 210 nm
injection
5 µL
sample/matrix
System suitability solution - 1.0 µg/mL of lamotrigine related compound B and 0.4 mg/ml of lamotrigine in methanol and buffer (60:40) v/v
Description
General description
Lamotrigine is an anti-convulsant medication used to treat epilepsy and to delay or prevent the recurrence of depressive episodes in bipolar disorder. For epilepsy, this includes focal seizures, tonic-clonic seizures, and seizures in Lennox-Gastaut syndrome.
In bipolar disorder, lamotrigine has not been shown to reliably treat acute depression, but for patients with bipolar disorder who are not currently symptomatic, it appears to be effective in reducing the risk of future episodes of depression.
This application illustrated the system suitability for the organic impurity analysis for lamotrigine tablets testing following the currently official United States Pharmacopoeia monograph
Analysis Note
For the lamotrigine standard solution, the RSD was 0.43 (USP specification NMT 10% RSD) and the tailing factor for lamotrigine was 1.7 (USP specification NMT 2). For the displayed system suitability solution the resolution between lamotrigine and the related compound B was 2.6 (USP specification NLT 2.0).
Other Notes
App_367I
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany