Merck
HPLC Analysis of Warfarin Enantiomers on Astec® CHIROBIOTIC® V (Method 1)
HPLC Analysis of Warfarin Enantiomers on Astec® CHIROBIOTIC® V (Method 1) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Astec® CHIROBIOTIC® V Chiral HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
Application
Product No.
Description
Pricing
HPLC Analysis of Warfarin Enantiomers on Astec® CHIROBIOTIC® V2
application for HPLC
HPLC Analysis of Warfarin Enantiomers on Astec® CHIROBIOTIC® V (Method 2)
application for HPLC
HPLC Analysis of Warfarin Enantiomers on Astec® Cellulose DMP
application for HPLC
HPLC Analysis of Warfarin Enantiomers on Astec® CYCLOBOND I 2000 (UV at 278 nm)
application for HPLC
standard
Product No.
Description
Pricing
Warfarin
analytical standard
Warfarin
PESTANAL®, analytical standard
(R)-(+)-Warfarin
≥97% (HPLC)
(S)-(−)-Warfarin
≥97% (HPLC)
CONDITIONS
column
CHIROBIOTIC V, 25 cm x 4.6 mm I.D., 5 μm particles (11024AST)
mobile phase
[A] acetonitrile; [B] 5 mM ammonium acetate, pH 4.1; (30:70, A:B)
flow rate
1 mL/min
column temp.
25 °C
detector
UV, 254 nm
injection
5 μL
sample
1 mg/mL in mobile phase
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC
null