Merck
HPLC Analysis of Triacylglycerols in Soybean Oil on Discovery® HS C18 (Acetone-Acetonitrile Mobile Phase, ELSD Detection)
HPLC Analysis of Triacylglycerols in Soybean Oil on Discovery® HS C18 (Acetone-Acetonitrile Mobile Phase, ELSD Detection) application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Discovery® HS C18 HPLC Column
5 μm particle size, L × I.D. 25 cm × 4.6 mm
standard
Product No.
Description
Pricing
Glyceryl trioleate
≥99%
Glyceryl trilinoleate
≥98% (TLC), liquid
Glyceryl trilinolenate
≥97% (TLC), liquid
Soybean oil
analytical standard
CONDITIONS
column
Discovery HS C18, 25 cm x 4.6 mm I.D., 5 μm particles (568523-U)
mobile phase
[A] acetone; [B] acetonitrile; (60:40, A:B)
flow rate
1.5 mL/min
pressure
914 psi (63 bar)
column temp.
30 °C
detector
ELSD
injection
5 μL
sample
5% soybean oil in acetone (w/v)
Description
Analysis Note
Soybean oil was analyzed as 5% diluted sample in acetone (w/v)
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany