Merck
HPLC Analysis of Pegylated Exenatide on Zenix® SEC-300 and Zenix®-C SEC-300
HPLC Analysis of Pegylated Exenatide on Zenix® SEC-300 and Zenix®-C SEC-300 application for HPLC
Materials
analytical column
Product No.
Description
Pricing
Zenix® SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steel
Zenix®-C SEC-300 Gel Filtration Column, 3 μm
L × I.D. 30 cm × 7.8 mm, 3 μm particle size, stainless steel
CONDITIONS
column
Zenix SEC-300, 30 cm x 7.8 mm I.D., 3 μm particles (Z777033)
column
Zenix-C SEC-300, 30 cm x 7.8 mm, I.D., 3 μm particles (Z777092)
mobile phase
(A) 50 mM Ammonium acetate: (B) acetonitrile; (90:10, A:B, v/v)
flow rate
0.5 mL/min
pressure
609 psi (42 bar)
column temp.
25 °C
detector
UV, 214 nm
injection
15 μL
sample
3.3 mg/ml PEG-Exanatide in water (PEG 23 kD)
Description
Legal Information
Zenix is a registered trademark of Sepax Technologies