HPLC Analysis of Carboxyamido Methylated BSA Tryptic Digest on Discovery® RP-Amide C16
![HPLC Analysis of Carboxyamido Methylated BSA Tryptic Digest on Discovery® RP-Amide C16 application for HPLC](/deepweb/assets/sigmaaldrich/product/images/354/378/4e69b77b-4aee-44a6-9bab-147214c6c7fb/800/4e69b77b-4aee-44a6-9bab-147214c6c7fb.jpg)
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Discovery RP-AmideC16, 25 cm × 4.6 mm I.D., 5 μm particles (505013)
mobile phase
[A] 0.1% TFA, 1% acetonitrile in water; [B] 0.1% TFA, 40% acetonitrile
gradient
0-100% B in 130 min
flow rate
1 mL/min
column temp.
35 °C
detector
UV, 215 nm
Description
Legal Information
Discovery is a registered trademark of Merck KGaA, Darmstadt, Germany