HPLC Analysis of Antipsychotic Drugs on Ascentis® Express RP-Amide
Materials
analytical column
CONDITIONS
column
Ascentis Express RP-Amide, 10 cm x 2.1 mm I.D., 2.7 μm particles (53913-U)
mobile phase
[A] 5 mM ammonium formate (in water); [B] 5 mM ammonium formate (in 95:5 methanol:water)
flow rate
3 mL/min
column temp.
35 °C
detector
ESI(+), TIC 100 - 1000 m/z
injection
1 μL
sample
200 μg/L in water
Description
Legal Information
Ascentis is a registered trademark of Merck KGaA, Darmstadt, Germany