HPLC Analysis of 3,4-Dihydroxyphenylalanine (DOPA) Enantiomers on Astec® CHIROBIOTIC® T
![HPLC Analysis of 3,4-Dihydroxyphenylalanine (DOPA) Enantiomers on Astec® CHIROBIOTIC® T application for HPLC](/deepweb/assets/sigmaaldrich/product/images/148/094/992989b1-0bd3-419c-b915-326e0671f025/800/992989b1-0bd3-419c-b915-326e0671f025.jpg)
CONDITIONS
column
CHIROBIOTIC T, 25 cm x 4.6 mm I.D., 5 μm particles (12024AST)
mobile phase
[A] water; [B] acetonitrile; (25:75, A:B)
flow rate
0.9 mL/min
column temp.
25 °C
detector
UV, 220 nm
injection
5 μL
sample
2 mg/mL in methanol
Description
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIROBIOTIC is a registered trademark of Sigma-Aldrich Co. LLC