Merck
GC Analysis of Wieland-Miescher Ketone Enantiomers on Astec® CHIRALDEX G-DP
GC Analysis of Wieland-Miescher Ketone Enantiomers on Astec® CHIRALDEX™ G-DP suitable for GC
Materials
standard
Product No.
Description
Pricing
(±)-8a-Methyl-3,4,8,8a-tetrahydro-1,6(2H,7H)-naphthalenedione
98%
CONDITIONS
column
Astec CHIRALDEX G-DP, 10 m x 0.25 mm I.D., 0.12 μm (78031AST)
oven
155 °C
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 12 psi
sample
peak 1: S(+)-Wieland-Miescher Ketone
peak 2: R(-)-Wieland-Miescher Ketone
Description
Analysis Note
Elution order reversal on CHIRALDEX G-BP.
Legal Information
Astec is a registered trademark of Merck KGaA, Darmstadt, Germany
CHIRALDEX is a trademark of Sigma-Aldrich Co. LLC