Merck
GC Analysis of Permanent Gases and Light Hydrocarbons on Supel-Q PLOT
GC Analysis of Permanent Gases and Light Hydrocarbons on Supel-Q PLOT suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Supel-Q PLOT Capillary GC Column
L × I.D. 30 m × 0.53 mm, average thickness 30 μm
CONDITIONS
column
Supel-Q PLOT, 30 m × 0.53 mm I.D. (25462)
oven
35 °C (3 min), 16 °C/min to 250 °C
inj. temp.
200 °C
detector
TCD, 250 °C
carrier gas
helium, 3 mL/min
injection
0.1 μL
sample
9-Component permanent gas a light hydrocarbon standard, each analyte at 1.0% in nitrogen
Description
Legal Information
null