GC Analysis of PUFA No. 3 Mix (Menhaden Oil Polyunsaturated Fatty Acid (PUFA) Methyl Esters) on Omegawax (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
Materials
analytical column
Product No.
Description
Pricing
CONDITIONS
column
Omegawax 100, 15 m × 0.10 mm I.D., 0.10 μm (23399-U)
oven
140 °C, 40 °C/min to 280 °C (2 min)
inj. temp.
250 °C
detector
FID, 280 °C
carrier gas
hydrogen, 50 cm/sec constant
injection
0.2 μL, 200:1 split
liner
4 mm I.D., split, cup design
sample
PUFA No. III – Menhaden Oil (47085-U), diluted to 50 mg/mL in methylene chloride
Description
Legal Information
null