Merck
GC Analysis of PUFA No. 2 Mix (Animal Source Polyunsatuated Fatty Acid (PUFA) Methyl Esters) on Omegawax (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of PUFA No. 2 Mix (Animal Source Polyunsatuated Fatty Acid (PUFA) Methyl Esters) on Omegawax (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Omegawax Capillary GC Column
L × I.D. 15 m × 0.10 mm, df 0.10 μm
CONDITIONS
column
Omegawax 100, 15 m × 0.10 mm I.D., 0.10 μm (23399-U)
oven
140 °C, 40 °C/min to 280 °C (2 min)
inj. temp.
250 °C
detector
FID, 280 °C
carrier gas
hydrogen, 50 cm/sec constant
injection
0.2 μL, 200:1 split
liner
4 mm I.D., split, cup design
sample
PUFA No. II – Animal Source (47015-U), diluted to 50 mg/mL in methylene chloride
Description
Legal Information
null