Skip to Content
End of year dates: Orders must be billed by 18.12. Merck Brazil will be closed from 20/12 to 06/01.
Merck
GC Analysis of Distilled Lime Essential Oil on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis
GC Analysis of Distilled Lime Essential Oil on Equity-1 (15 m x 0.10 mm I.D., 0.10 μm), Fast GC Analysis suitable for GC
Materials
analytical column
Product No.
Description
Pricing
Equity-1 Capillary GC Column
L × I.D. 15 m × 0.10 mm, df 0.10 μm
CONDITIONS
column
Equity-1, 15 m × 0.10 mm I.D., 0.10 μm (28039-U)
oven
75 °C (1 min), 35 °C/min to 200 °C (1 min)
inj. temp.
250 °C
detector
FID, 250 °C
carrier gas
helium, 45 cm/sec constant
injection
0.10 μL, 300:1 split
liner
2 mm I.D., straight
sample
distilled lime oil, neat
Description
Legal Information
null