- CD49d expression is an independent risk factor of progressive disease in early stage chronic lymphocytic leukemia.
CD49d expression is an independent risk factor of progressive disease in early stage chronic lymphocytic leukemia.
Identification of prognosticators for Binet A chronic lymphocytic leukemia is important for selecting patients with dismal prognosis. We analyzed CD49d expression in 140 consecutive Binet A chronic lymphocytic leukemia. At diagnosis, CD49d >or=30% (54/140, 38.6%) associated with proliferation markers, namely CD38 >or=30% (p=3.9 x 10(-6)), LDH (p=0.007) and beta2-microglobulin (p=0.020). Univariate log-rank analysis identified CD49d >or=30% as a risk factor of treatment free survival (p=8.3 x 10(-5)), time to progression to a more advanced stage (p=4.7 x 10(-4)), and time to lymphocyte doubling (p=0.009). Multivariate analysis selected CD49d >or=30% as an independent treatment free survival predictor after adjustment for biological (HR 2.28; 95% CI 1.71-4.45, p=0.015) and both biological and clinical variables analyzed together (HR 3.33, 95% CI 1.61-6.90, p=0.001). Within Binet A subgroups harboring favorable biological variables (IGHV homology <98%, favorable karyotype, CD38 <30%, ZAP70 <20%) or clinical variables, CD49d >or=30% consistently identified a subset of patients with short treatment free survival. Our observations indicate CD49d >or=30% as a new marker for the initial prognostic assessment of Binet A chronic lymphocytic leukemia.