Merck
  • Phosphatidylinositol-4-phosphate 5-kinases and phosphatidylinositol 4,5-bisphosphate synthesis in the brain.
Phosphatidylinositol-4-phosphate 5-kinases and phosphatidylinositol 4,5-bisphosphate synthesis in the brain.
The Journal of biological chemistry (2010-07-14)
Laura A Volpicelli-Daley, Louise Lucast, Liang-Wei Gong, Lijuan Liu, Junko Sasaki, Takehiko Sasaki, Charles S Abrams, Yasunori Kanaho, Pietro De Camilli
ABSTRACT
The predominant pathway for phosphatidylinositol (4,5)-bisphosphate (PI(4,5)P(2)) synthesis is thought to be phosphorylation of phosphatidylinositol 4-phosphate at the 5 position of the inositol ring by type I phosphatidylinositol phosphate kinases (PIPK): PIPKIalpha, PIPKIbeta, and PIPKIgamma. PIPKIgamma has been shown to play a role in PI(4,5)P(2) synthesis in brain, and the absence of PIPKIgamma is incompatible with postnatal life. Conversely, mice lacking PIPKIalpha or PIPKIbeta (isoforms are referred to according to the nomenclature of human PIPKIs) live to adulthood, although functional effects in specific cell types are observed. To determine the contribution of PIPKIalpha and PIPKIbeta to PI(4,5)P(2) synthesis in brain, we investigated the impact of disrupting multiple PIPKI genes. Our results show that a single allele of PIPKIgamma, in the absence of both PIPKIalpha and PIPKIbeta, can support life to adulthood. In addition, PIPKIalpha alone, but not PIPKIbeta alone, can support prenatal development, indicating an essential and partially overlapping function of PIPKIalpha and PIPKIgamma during embryogenesis. This is consistent with early embryonic expression of PIPKIalpha and PIPKIgamma but not of PIPKIbeta. PIPKIbeta expression in brain correlates with neuronal differentiation. The absence of PIPKIbeta does not impact embryonic development in the PIPKIgamma knock-out (KO) background but worsens the early postnatal phenotype of the PIPKIgamma KO (death occurs within minutes rather than hours). Analysis of PIP(2) in brain reveals that only the absence of PIPKIgamma significantly impacts its levels. Collectively, our results provide new evidence for the dominant importance of PIPKIgamma in mammals and imply that PIPKIalpha and PIPKIbeta function in the generation of specific PI(4,5)P(2) pools that, at least in brain, do not have a major impact on overall PI(4,5)P(2) levels.
Sigma-Aldrich
Phosphoinositides sodium salt from bovine brain
Pricing and availability is not currently available.