Merck
  • Polynuclear aromatic hydrocarbon and particulate emissions from two-stage combustion of polystyrene: the effects of the secondary furnace (afterburner) temperature and soot filtration.
Polynuclear aromatic hydrocarbon and particulate emissions from two-stage combustion of polystyrene: the effects of the secondary furnace (afterburner) temperature and soot filtration.
Environmental science & technology (2002-03-07)
Jun Wang, Henning Richter, Jack B Howard, Yiannis A Levendis, Joel Carlson
ABSTRACT
Laboratory experiments were conducted in a two-stage horizontal muffle furnace in order to monitor emissions from batch combustion of polystyrene (PS) and identify conditions that minimize them. PS is a dominant component of municipal and hospital waste streams. Bench-scale combustion of small samples (0.5 g) of shredded styrofoam cups was conducted in air, using an electrically heated horizontal muffle furnace, kept at Tgas = 1000 degrees C. Upon devolatilization, combustion of the polymer took place in a diffusion flame over the sample. The gaseous combustion products were mixed with additional air in a venturi and were channeled to a secondary muffle furnace (afterburner) kept at Tgas = 900-1100 degrees C; residence time therein varied between 0.6 and 0.8 s. At the exits of the primary and the secondary furnace the emissions of CO, CO2, O2, NOx, particulates as well as volatile and semivolatile hydrocarbons, such as polycyclic aromatic hydrocarbons (PAH), were monitored. Online analyzers, gravimetric techniques, and gas chromatography coupled to mass spectrometry (GC-MS) were used. Experiments were also conducted with a high-temperature barrier filter, placed just before the exit of the primary furnace to prevent the particulates from entering into the secondary furnace. Results demonstrated the beneficial effect of the afterburner in reducing PAH concentrations, including those of mutagenic species such as benzo[a]pyrene. Concentrations of individual PAH exhibited a pronounced after burner temperature dependence, typically ranging from a small decrease at 900 degrees C to a larger degree of consumption at 1100 degrees C. Consumption of PAH was observed to be the dominant feature at 900 degrees C, while significant quantities of benzene and some of its derivatives, captured by means of carbosieve/Carbotrap adsorbents, were formed in the afterburner at a temperature of 1000 degrees C. In the primary furnace, about 30% of the mass of the initial polystyrene was converted into soot, while the total mass of PAH represented about 3% of the initial mass of combustible. The afterburner reduced the particulate (soot) emissions by only 20-30%, which indicates that once soot is formed its destruction is rather difficult because its oxidation kinetics are slow undertypical furnace conditions. Moreover, increasing the afterburnertemperature resulted in an increasing trend of soot emissions therefrom, which might indicate competition between soot oxidation and formation, with some additional formation occurring at the higher temperatures. Contrary to the limited effect of the afterburner, high-temperature filtration of the combustion effluent prior to the exit of the primary furnace allowed for effective soot oxidation inside of the ceramic filter. Filtration drastically reduced soot emissions, by more than 90%. Limited soot formation in the afterburner was again observed with increasing temperatures. The yields of both CO and CO2 were largely unaffected by the temperature of the afterburner but increased at the presence of the filter indicating oxidation therein. A previously developed kinetic model was used to identify major chemical reaction pathways involving PAH in the afterburner. The experimental data at the exit of the primary furnace was used as input to these model computations. A first evaluation of the predictive capability of the model was conducted for the case with ceramic filter and a temperature of 900 degrees C. The afterburner was approximated as a plug-flow reactor, and model predictions at a residence time of 0.8 s were compared to experimental data collected at its exit. In agreement with the experimental PAH concentration, only a minor impact of the afterburner treatment was observed for most species at 900 degrees C. OH was deduced to be the major reactant with a mole fraction about 4 orders of magnitudes higher than that of hydrogen radicals. Evidence for the need of further work on the quantitative assessment of oxidation of PAH and their radicals is given.
Product Description
Supelco
Carbotrap® 300, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbotrap® 349, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbotrap® 300, stainless steel TD tube, O.D. × I.D. × L 1/4 in. × 5 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® C, 20-40 mesh, bottle of 10 g
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® B, 20-40 mesh, bottle of 10 g
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® X, 20-40 mesh, bottle of 10 g
Supelco
Carbotrap® Adsorbent, matrix Carbotrap® C, 20-40 mesh, bottle of 500 g
Supelco
Carbotrap® 300, stainless steel TD tube, unconditioned, pkg of 10 ea
Supelco
Carbotrap® 300, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, preconditioned, pkg of 10 ea
Supelco
Carbotrap® 300, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbotrap® 300, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbotrap® 300, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbotrap® 349
Supelco
Carbotrap® 202, stainless steel TD tube, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
ORBO 101 Carbotrap® B (20/40), 100/50 mg, W,W,W separators, O.D. × L 6 mm × 70 mm, pkg of 25 ea
Supelco
ORBO 100 on Carbotrap® B (20/40), 350/175 mg, W,W,W separators, O.D. × L 7 mm × 110 mm, pkg of 25 ea
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 60-80 mesh, bottle of 5 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 80-100 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve G, 80-100 mesh, bottle of 5 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-III, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve Adsorbent, matrix Carbosieve S-II, 60-80 mesh, bottle of 10 g
Supelco
Carbosieve SIII, stainless steel TD tube, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbotrap® 217, glass TD tube, fritted, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4 × 3 1/2, unconditioned, pkg of 10 ea
Supelco
Carbotrap® 202, glass TD tube, fritted, O.D. × I.D. × L 6 mm × 4 mm × 7 in., preconditioned, pkg of 1 ea
Supelco
Carbotrap® 217, stainless steel TD tube, O.D. × L 1/4 in. × 3 1/2 in., preconditioned, pkg of 10 ea
Supelco
Carbotrap® 217, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbotrap® 217, O.D. × L 1/4 in. × 3 1/2 in., glass TD tube, fritted, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea
Supelco
Carbotrap® 202, O.D. × L 1/4 in. × 3 1/2 in., stainless steel TD tube, Sealed with Brass Endcaps, preconditioned, pkg of 10 ea