Merck
  • A comparative study of the reversed-phase HPLC retention behaviour of S-adenosyl-L-methionine and its related metabolites on Hypersil ODS and Supelcosil LC-ABZ stationary phases.
A comparative study of the reversed-phase HPLC retention behaviour of S-adenosyl-L-methionine and its related metabolites on Hypersil ODS and Supelcosil LC-ABZ stationary phases.
Journal of pharmaceutical and biomedical analysis (1993-04-01)
K Valkó, M P Hamedani, T L Ascah, W A Gibbons
ABSTRACT
S-Adenosyl-L-methionine (SAM) and its metabolites S-adenosyl-L-homocysteine (SAH) and methyl-thioadenosine (MTA) are endogenous compounds that are heavily involved in a variety of biochemical processes, and have therefore been the target for several assays in body fluids and tissues. Reversed-phase chromatographic behaviour of SAM and its metabolites has been studied by using Supelcosil LC-ABZ column, specially designed for analysis of acidic, basic, zwitterionic and neutral compounds, and on a Hypersil ODS column as a function of mobile phase pH. The retentions of the compounds, expressed by the capacity ratio (k'), are measured on both column with mobile phases comprised of 10% acetonitrile and 10 mM ammonium formate buffer with pH values ranging from 2 to 9. Higher selectivity is observed on Supelcosil LC-ABZ within pH range 4-6. Different retention properties are observed at very low pH and seemed as if the Supelcosil LC-ABZ column reduced the effect of the mobile phase pH by about 1 pH unit. Whilst the Supelcosil column can be recommended for the routine analysis of SAM and its related metabolites in biological fluids by using mobile phase pH 5, the Hypersil ODS column may be suggested for use with mobile phase pH values of 3-4.
Product Description
Supelco
SUPELCOSIL LC-NH2-NP HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Pricing and availability is not currently available.
Supelco
SUPELCOSIL LC-NH2-NP Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Pricing and availability is not currently available.
Supelco
SUPELCOSIL LC-ABZ Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Pricing and availability is not currently available.
Supelco
SUPELCOSIL LC-ABZ Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Pricing and availability is not currently available.
Supelco
SUPELCOSIL LC-ABZ Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 4 mm
Pricing and availability is not currently available.
Supelco
SUPELCOSIL LC-ABZ HPLC Column, 5 μm particle size, L × I.D. 5 cm × 4.6 mm
Pricing and availability is not currently available.
Supelco
SUPELCOSIL LC-ABZ HPLC Column, 5 μm particle size, L × I.D. 25 cm × 3 mm
Pricing and availability is not currently available.
Supelco
SUPELCOSIL LC-ABZ HPLC Column, 5 μm particle size, L × I.D. 15 cm × 4.6 mm
Pricing and availability is not currently available.
Supelco
SUPELCOSIL LC-ABZ HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4.6 mm
Pricing and availability is not currently available.
Supelco
SUPELCOSIL LC-ABZ HPLC Column, 5 μm particle size, L × I.D. 25 cm × 4 mm
Pricing and availability is not currently available.
Supelco
SUPELCOSIL LC-ABZ Supelguard Cartridge, 5 μm particle size, L × I.D. 2 cm × 2.1 mm
Pricing and availability is not currently available.