Merck
  • Method optimization and validation for the determination of eight sulfonamides in chicken muscle and eggs by modified QuEChERS and liquid chromatography with fluorescence detection.
Method optimization and validation for the determination of eight sulfonamides in chicken muscle and eggs by modified QuEChERS and liquid chromatography with fluorescence detection.
Journal of pharmaceutical and biomedical analysis (2016-03-13)
José Fernando Huertas-Pérez, Natalia Arroyo-Manzanares, Lucie Havlíková, Laura Gámiz-Gracia, Petr Solich, Ana M García-Campaña
ABSTRACT
A simple, effective and reliable method for the determination of eight sulfonamide antibiotics (sulfadiazine, sulfapiridine, sulfamerazine, sulfamethazine, sulfachloropiridazine, sulfamethoxazole, sulfadoxine, sulfadimethoxin) in chicken muscle and eggs by liquid chromatography and fluorescence detection has been developed and validated. Sulfonamides do not present native fluorescence, however their direct determination was achieved by on-line post-column photochemical derivatization by UV irradiation. Sample treatment was based on QuEChERS with several modifications depending on the matrix. Egg extracts were cleaned-up using PSA for the dispersive solid phase extraction step. On the other hand, a new clean-up sorbent, Supel™ QuE Z-Sep(+), has been successfully applied in chicken muscle extract and has proved to be effective for interference removal from this matrix. Under optimum conditions, recoveries from 65.9 to 88.1%, relative standard deviations lower than 10% (except for sulfachloropiridazine), and limits of quantification (LOQs) from 14 to 85 μg kg(-1) were achieved. Thus, the method complies with current European requirements.
Product Description
Supelco
Supel QuE, Z-Sep+ Bulk, pkg of 20 g
Supelco
Supel QuE, Z-Sep/C18 Tube, centrifuge tube volume 2 mL, pack of 100
Supelco
Supel QuE, Z-Sep+ Tube, centrifuge tube volume 2 mL, pack of 100 ea
Supelco
Supel QuE, Z-Sep+ Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep/C18 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep Bulk
Supelco
Supel QuE, Z-Sep+ with MgSO4 Tube, pk of 50, centrifuge tube volume 15 mL , Shaker Compatible
Supelco
Supel QuE, Z-Sep Tube, centrifuge tube volume 2 mL, pack of 100 ea