Merck
Collagen Type 1 Accelerates Healing of Ruptured Fetal Membranes.
Scientific reports (2018-01-14)
Haruta Mogami, Annavarapu Hari Kishore, R Ann Word
ABSTRACT
Preterm premature rupture of membranes (pPROM) is a major cause of preterm birth. Recently, extracellular matrix-directed treatment is applied for wound healing. Here, we used a pregnant mouse model to test the efficacy of collagen type 1 gel for healing of the prematurely ruptured fetal membranes. Although injection of PBS into the ruptured fetal membranes resulted in 40% closure, injection of collagen type 1 improved closure rates to 90% within 72 h. Macrophages of the M2 wound healing phenotype were entrapped in the collagen layer. In primary human amnion mesenchymal cells, collagen type 1 gels activated collagen receptor discoidin domain receptor 2 (DDR2) to induce myosin light chain phosphorylation and migration of injured amnion mesenchymal cells. These findings define the mechanisms for matrix-directed therapeutics for pPROM.
Sigma-Aldrich
Dulbecco′s Modified Eagle′s Medium/Nutrient Mixture F-12 Ham, With L-glutamine, 15 mM HEPES, and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Screen cup for CD-1, size 85 mL
Pricing and availability is not currently available.
Sigma-Aldrich
DDR1-IN-1, ≥98% (HPLC)
Pricing and availability is not currently available.
Sigma-Aldrich
O-Phospho-L-tyrosine
Pricing and availability is not currently available.
Sigma-Aldrich
Screens for CD-1, size 40 mesh
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Integrin α2 Antibody, clone P1E6, azide free, clone P1E6, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Integrin β1 Antibody, clone 6S6, Azide Free, clone 6S6, Chemicon®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Phosphotyrosine Antibody, clone 4G10®, clone 4G10®, Upstate®, from mouse
Pricing and availability is not currently available.
Sigma-Aldrich
Anti-Integrin α1 Antibody, clone FB12, azide free, clone FB12, Chemicon®, from mouse
Pricing and availability is not currently available.