Merck
  • Specific cyclic ADP-ribose phosphohydrolase obtained by mutagenic engineering of Mn
Specific cyclic ADP-ribose phosphohydrolase obtained by mutagenic engineering of Mn
Scientific reports (2018-01-20)
João Meireles Ribeiro, José Canales, Alicia Cabezas, Joaquim Rui Rodrigues, Rosa María Pinto, Iralis López-Villamizar, María Jesús Costas, José Carlos Cameselle
ABSTRACT
Cyclic ADP-ribose (cADPR) is a messenger for Ca
Sigma-Aldrich
β-Nicotinamide mononucleotide, ≥95% (HPLC)