Merck
  • Mechanism of Antiwear Property Under High Pressure of Synthetic Oil-Soluble Ultrathin MoS
Mechanism of Antiwear Property Under High Pressure of Synthetic Oil-Soluble Ultrathin MoS
Langmuir : the ACS journal of surfaces and colloids (2018-01-02)
Zhe Chen, Yuhong Liu, Selda Gunsel, Jianbin Luo
ABSTRACT
Wear occurs between two rubbing surfaces. Severe wear due to seizure under high pressure leads to catastrophic failures of mechanical systems and raises wide concerns. In this paper, a kind of synthetic oil-soluble ultrathin MoS
Sigma-Aldrich
Ammonium tetrathiomolybdate, 99.97% trace metals basis