Merck
Store-operated Ca
JCI insight (2017-03-30)
Miriam Eckstein, Martin Vaeth, Cinzia Fornai, Manikandan Vinu, Timothy G Bromage, Meerim K Nurbaeva, Jessica L Sorge, Paulo G Coelho, Youssef Idaghdour, Stefan Feske, Rodrigo S Lacruz
ABSTRACT
Loss-of-function mutations in stromal interaction molecule 1 (STIM1) impair the activation of Ca
Sigma-Aldrich
Anti-STIM1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution
Pricing and availability is not currently available.