Merck
  • Inhibition of PKCα reduces the ability of migration of kidney cancer cells but has no impact on cell apoptosis.
Inhibition of PKCα reduces the ability of migration of kidney cancer cells but has no impact on cell apoptosis.
Experimental and therapeutic medicine (2017-06-02)
Bo Zhan, Chuize Kong, Zhe Zhang, Xiao Dong, Naiwen Zhang
ABSTRACT
Kidney cancer is among the most important causes of cancer-associated mortality worldwide. The present study aimed to evaluate protein kinase C α (PKCα) expression in kidney cancer tissues and cell lines, and its significance in apoptosis and migration. Expression of PKCα was analyzed using quantitative polymerase chain reaction and western blotting. In addition, the inhibitor of PKCα (calphostin C and GO6976) was used to treat kidney cancer cells. The ACHN cell line was generated with PKCα-small-interfering RNA (siRNA) and a stable expression of PKCα, in order to facilitate the analysis of apoptosis and migration of PKCα during knockdown and inactivation. Flow cytometry was used to determine the rates of apoptosis. Immunohistochemical staining was used to identify the localization of PKCα in renal clear cell carcinoma and normal sections. PKCα expression in normal tissues was found to be greater than in cancerous tissues. Furthermore, apoptosis was not promoted with PKCα inhibitors or PKCα-siRNA treatment, and a decrease of the migration ability was observed following transfection with PKCα-dominant negative. The results indicated that inhibition of PKCα might not contribute to apoptosis progression in kidney carcinoma.
Sigma-Aldrich
Monoclonal Anti-GAPDH antibody produced in mouse, clone 3C2, purified immunoglobulin, buffered aqueous solution
Pricing and availability is not currently available.