Merck
  • Disordered clusters of Bak dimers rupture mitochondria during apoptosis.
Disordered clusters of Bak dimers rupture mitochondria during apoptosis.
eLife (2017-02-10)
Rachel T Uren, Martin O'Hely, Sweta Iyer, Ray Bartolo, Melissa X Shi, Jason M Brouwer, Amber E Alsop, Grant Dewson, Ruth M Kluck
ABSTRACT
During apoptosis, Bak and Bax undergo major conformational change and form symmetric dimers that coalesce to perforate the mitochondrial outer membrane via an unknown mechanism. We have employed cysteine labelling and linkage analysis to the full length of Bak in mitochondria. This comprehensive survey showed that in each Bak dimer the N-termini are fully solvent-exposed and mobile, the core is highly structured, and the C-termini are flexible but restrained by their contact with the membrane. Dimer-dimer interactions were more labile than the BH3:groove interaction within dimers, suggesting there is no extensive protein interface between dimers. In addition, linkage in the mobile Bak N-terminus (V61C) specifically quantified association between dimers, allowing mathematical simulations of dimer arrangement. Together, our data show that Bak dimers form disordered clusters to generate lipidic pores. These findings provide a molecular explanation for the observed structural heterogeneity of the apoptotic pore.
Sigma-Aldrich
Anti-Bak antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Pricing and availability is not currently available.