- Lumican delays melanoma growth in mice and drives tumor molecular assembly as well as response to matrix-targeted TAX2 therapeutic peptide.
Lumican delays melanoma growth in mice and drives tumor molecular assembly as well as response to matrix-targeted TAX2 therapeutic peptide.
Scientific reports (2017-08-11)
Albin Jeanne, Valérie Untereiner, Corinne Perreau, Isabelle Proult, Cyril Gobinet, Camille Boulagnon-Rombi, Christine Terryn, Laurent Martiny, Stéphane Brézillon, Stéphane Dedieu
PMID28794454
ABSTRACT
Lumican is a small leucine-rich proteoglycan (SLRP) being known as a key regulator of collagen fibrillogenesis. However, little attention has been given so far in studying its influence on tumor-associated matrix architecture. Here, we investigate the role of host lumican on tumor matrix organization as well as on disease progression considering an immunocompetent model of melanoma implanted in Lum
Sigma-Aldrich
Anti-LUM antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution