Merck
  • Role of 6-O-α-maltosyl-β-cyclodextrin in lysosomal cholesterol deprivation in Npc1-deficient Chinese hamster ovary cells.
Role of 6-O-α-maltosyl-β-cyclodextrin in lysosomal cholesterol deprivation in Npc1-deficient Chinese hamster ovary cells.
Carbohydrate research (2017-11-25)
Yasuyo Okada, Erika Ueda, Yuki Kondo, Yoichi Ishitsuka, Tetsumi Irie, Taishi Higashi, Keiichi Motoyama, Hidetoshi Arima, Muneaki Matuso, Katsumi Higaki, Kousaku Ohno, Junichi Nishikawa, Atsushi Ichikawa
ABSTRACT
We aimed to investigate whether 6-O-α-maltosyl-β-cyclodextrin (Mal-βCD) is incorporated into cells and lysosomes during the release of unesterified cholesterol in Npc1-deficient Chinese hamster ovary (CHO) cells (Npc1 KO cells) and CHO-JP17 cells (JP17 cells). Internalization of Mal-βCD in cells and lysosomes and extracellular release of lysosomal unesterified cholesterol were demonstrated by LC/MS/MS and LC/MS, respectively. Internalization of Mal-βCD was greater in Npc1 KO cells than in JP17 cells. The majority of internalized Mal-βCD in both cell types was metabolized by lysosomal α-glucosidase to 6-O-α-D-glucosyl-β-cyclodextrin (Glc-βCD). However, Mal-βCD did not directly enter the lysosomes prepared from cell homogenates. Mal-βCD-treated Npc1 KO cells and JP17 cells both released Mal-βCD and Glc-βCD, together with unesterified cholesterol, out of cells. The release of unesterified cholesterol by Mal-βCD in Npc1 KO cells was much greater than that in JP17 cells. This study is the first to report the influx of Mal-βCD into the lysosomes of Npc1 KO cells and JP17 cells, followed by generation of Glc-βCD, and the efflux of Mal-βCD/Glc-βCD and unesterified cholesterol to the extracellular fluid, based on the quantitative LC/MS analysis.
Sigma-Aldrich
Cholesterol-2,2,3,4,4,6-d6, 97 atom % D, 98% (CP)