Merck
  • Neuron-derived transthyretin modulates astrocytic glycolysis in hormone-independent manner.
Neuron-derived transthyretin modulates astrocytic glycolysis in hormone-independent manner.
Oncotarget (2018-01-02)
Alina Zawiślak, Piotr Jakimowicz, James A McCubrey, Dariusz Rakus
ABSTRACT
It has been shown that neurons alter the expression of astrocytic metabolic enzymes by secretion of until now unknown molecule(s) into extracellular fluid. Here, we present evidence that neuron-derived transthyretin (TTR) stimulates expression of glycolytic enzymes in astrocytes which is reflected by an increased synthesis of ATP. The action of TTR is restricted to regulatory enzymes of glycolysis: phosphofructokinase P (PFKP) and pyruvate kinase M1/M2 isoforms (PKM1/2). The regulation of PFK and PKM expression by TTR is presumably specific for brain tissue and is independent of the role of TTR as a carrier protein for thyroxine and retinol. TTR induced expression of PKM and PFK is mediated by the cAMP/PKA-dependent pathway and is antagonized by the PI3K/Akt pathway. Our results provide the first experimental evidence for action of TTR as a neuron-derived energy metabolism activator in astrocytes and describe the mechanisms of its action. The data presented here suggest that TTR is involved in a mechanism in which neurons stimulate degradation of glycogen-derived glucosyl units without significant modulation of glucose uptake by glial cells.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-Mouse IgG (whole molecule)–Peroxidase antibody produced in goat, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–FITC antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
Anti-Glial Fibrillary Acidic Protein antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution
Sigma-Aldrich
ExtrAvidin®−Peroxidase, buffered aqueous solution
Sigma-Aldrich
Anti-Goat IgG (whole molecule)–Biotin antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution