Merck
Nanostructured Pluronic hydrogels as bioinks for 3D bioprinting.
Biofabrication (2015-08-12)
Michael Müller, Jana Becher, Matthias Schnabelrauch, Marcy Zenobi-Wong
ABSTRACT
Bioprinting is an emerging technology in the field of tissue engineering as it allows the precise positioning of biologically relevant materials in 3D, which more resembles the native tissue in our body than current homogenous, bulk approaches. There is however a lack of materials to be used with this technology and materials such as the block copolymer Pluronic have good printing properties but do not allow long-term cell culture. Here we present an approach called nanostructuring to increase the biocompatibility of Pluronic gels at printable concentrations. By mixing acrylated with unmodified Pluronic F127 it was possible to maintain the excellent printing properties of Pluronic and to create stable gels via UV crosslinking. By subsequent elution of the unmodified Pluronic from the crosslinked network we were able to increase the cell viability of encapsulated chondrocytes at day 14 from 62% for a pure acrylated Pluronic hydrogel to 86% for a nanostructured hydrogel. The mixed Pluronic gels also showed good printability when cells where included in the bioink. The nanostructured gels were, with a compressive modulus of 1.42 kPa, mechanically weak, but we were able to increase the mechanical properties by the addition of methacrylated hyaluronic acid. Our nanostructuring approach enables Pluronic hydrogels to have the desired set of properties in all stages of the bioprinting process.
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) diacrylate, average Mn ~14,600
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) diacrylate, average Mn ~5,800
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) dimethacrylate, average Mn ~14,600
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) dimethacrylate, average Mn ~12,500
Sigma-Aldrich
Poly(ethylene glycol)-block-poly(propylene glycol)-block-poly(ethylene glycol) diacrylate, average Mn ~12,500