Merck
  • Tuning the Composition and Structure of Amorphous Molybdenum Sulfide/Carbon Black Nanocomposites by Radiation Technique for Highly Efficient Hydrogen Evolution.
Tuning the Composition and Structure of Amorphous Molybdenum Sulfide/Carbon Black Nanocomposites by Radiation Technique for Highly Efficient Hydrogen Evolution.
Scientific reports (2017-11-24)
Pengfei Cao, Jing Peng, Siqi Liu, Yu Cui, Yang Hu, Bo Chen, Jiuqiang Li, Maolin Zhai
ABSTRACT
Amorphous molybdenum sulfide/carbon black (MoS
Sigma-Aldrich
Ammonium tetrathiomolybdate, 99.97% trace metals basis