Merck
  • Significant enhanced uranyl ions extraction efficiency with phosphoramidate-functionalized ionic liquids via synergistic effect of coordination and hydrogen bond.
Significant enhanced uranyl ions extraction efficiency with phosphoramidate-functionalized ionic liquids via synergistic effect of coordination and hydrogen bond.
Scientific reports (2017-11-18)
Xiang Xie, Zhen Qin, Yao He, Penghui Xiong, Zeng Huang, Yiwu Mao, Hongyuan Wei, Liangang Zhuo
ABSTRACT
The influence of the linking group between the phosphoryl and bridging moieties in phosphoryl-containing task-specific ionic liquids (TSILs) on the extraction of uranyl ions was experimentally and theoretically investigated. A novel phosphoramidate-based TSIL with an amine group as the linking moiety resulted in a higher uranyl ion extraction efficiency compared with that of other phosphoryl-based TSILs. A distribution ratio of 4999 ± 51 can be achieved for uranyl ions. The uranyl ions (76.7 ± 1.5%) were stripped from the loaded ionic liquid phase in a single stage using 0.05 M diethylenetriamine pentaacetic acid in a 1.0 M guanidine carbonate solution. The extraction stoichiometry of the uranyl ions was determined by a slope analysis of the extraction data. Furthermore, the fundamental nature of the interaction between the phosphoramidate-based TSIL and uranyl ions was theoretically studied for the first time. The theoretical calculations demonstrated that the synergistic effect of the complexation interaction and H-bond formation between the phosphoramidate-functional ionic liquid and uranyl nitrate led to the higher extraction efficiency. These results provide a basis for rational design, synthesis and potential applications of novel TSILs for uranyl extraction.
Sigma-Aldrich
4-Bromo-1-butanol, technical, ≥85% (as solvent-free, GC)
Sigma-Aldrich
Dibutyl phosphate, ≥97.0% (T)