Merck
  • Phase behavior, rheological characteristics and microstructure of sodium caseinate-Persian gum system.
Phase behavior, rheological characteristics and microstructure of sodium caseinate-Persian gum system.
Carbohydrate polymers (2017-11-08)
Farzad Sadeghi, Rassoul Kadkhodaee, Bahareh Emadzadeh, Glyn O Phillips
ABSTRACT
In this study, the phase behavior of sodium caseinate-Persian gum mixtures was investigated. The effect of thermodynamic incompatibility on phase distribution of sodium caseinate fractions as well as the flow behavior and microstructure of the biopolymer mixtures were also studied. The phase diagram clearly demonstrated the dominant effect of Persian gum on the incompatibility of the two biopolymers. SDS-PAGE electrophoresis indicated no selective fractionation of sodium caseinate subunits between equilibrium phases upon de-mixing. The microstructure of mixtures significantly changed depending on their position within the phase diagram. Fitting viscometric data to Cross and Bingham models revealed that the apparent viscosity, relaxation time and shear thinning behavior of the mixtures is greatly influenced by the volume ratio and concentration of the equilibrium phases. There is a strong dependence of the flow behavior of sodium caseinate-Persian gum mixtures on the composition of the equilibrium phases and the corresponding microstructure of the system.
Sigma-Aldrich
Casein sodium salt from bovine milk
Pricing and availability is not currently available.