- Kastle-Meyer blood test reagents are deleterious to DNA.
Kastle-Meyer blood test reagents are deleterious to DNA.
Forensic science international (2017-11-13)
James Sloots, Wendy Lalonde, Barbara Reid, Jonathan Millman
PMID29128654
ABSTRACT
The Kastle-Meyer (KM) test is a quick and easy chemical test for blood used in forensic analyses. Two practical variations of this test are the KM-rub (indirect) test and the more sensitive KM-direct test, the latter of which is performed by applying reagents directly to a suspected blood stain. This study found that sodium hydroxide present in the KM reagents eliminated the potential to generate a DNA profile when applied directly to small quantities of blood. A modified approach to the KM-rub test that increases its sensitivity is presented as a method to replace destructive KM-direct testing.