Merck
  • Supramolecular gels: using an amide-functionalized imidazolium-based surfactant.
Supramolecular gels: using an amide-functionalized imidazolium-based surfactant.
Journal of colloid and interface science (2017-10-14)
Ni Cheng, Qi Kang, Jianhong Xiao, Na Du, Li Yu
ABSTRACT
Studies on supramolecular gel aggregates via supramolecular interactions are very fascinating and impressive. The interactions of gelator with solvent and inorganic salt can functionalize, activate, or control the properties of supramolecular gels. A lot of work on this area has been done, but all these contributions are only at an early stage. In this work, we tuned the gel formation conditions and properties by using low-molecular-weight gelator (LMWGs), amide-functionalized imidazolium-based surfactant, viz., N-cetyl-N'-acetamido imidazolium bromide ([N-C
Sigma-Aldrich
2-Bromoacetamide, 98%
Pricing and availability is not currently available.
Sigma-Aldrich
1-Bromohexadecane, 97%
Pricing and availability is not currently available.