Merck
  • Employing Calcination as a Facile Strategy to Reduce the Cytotoxicity in CoFe
Employing Calcination as a Facile Strategy to Reduce the Cytotoxicity in CoFe
ACS applied materials & interfaces (2017-10-24)
Débora R Lima, Ning Jiang, Xin Liu, Jiale Wang, Valcinir A S Vulcani, Alessandro Martins, Douglas S Machado, Richard Landers, Pedro H C Camargo, Alexandre Pancotti
ABSTRACT
CoFe