Merck
  • pH-Driven Wetting Switchability of Electrodeposited Superhydrophobic Copolymers of Pyrene Bearing Acid Functions and Fluorinated Chains.
pH-Driven Wetting Switchability of Electrodeposited Superhydrophobic Copolymers of Pyrene Bearing Acid Functions and Fluorinated Chains.
Chemphyschem : a European journal of chemical physics and physical chemistry (2017-09-01)
Gabriela Ramos Chagas, Denis Kiryanenko, Guilhem Godeau, Frédéric Guittard, Thierry Darmanin
ABSTRACT
A smart stimuli-responsive surface was fabricated by the electro-copolymerization of pyrene monomers followed by base and acid treatment. Copolymers of pyrenes bearing fluorinated chains (Py-nF
Sigma-Aldrich
1-Pyreneacetic acid, 97%
Pricing and availability is not currently available.