Merck
  • PSMA ligand conjugated PCL-PEG polymeric micelles targeted to prostate cancer cells.
PSMA ligand conjugated PCL-PEG polymeric micelles targeted to prostate cancer cells.
PloS one (2014-11-12)
Jian Jin, Bowen Sui, Jingxin Gou, Jingshuo Liu, Xing Tang, Hui Xu, Yu Zhang, Xiangqun Jin
ABSTRACT
In this content, a small molecular ligand of prostate specific membrane antigen (SMLP) conjugated poly (caprolactone) (PCL)-b-poly (ethylene glycol) (PEG) copolymers with different block lengths were synthesized to construct a satisfactory drug delivery system. Four different docetaxel-loaded polymeric micelles (DTX-PMs) were prepared by dialysis with particle sizes less than 60 nm as characterized by dynamic light scattering (DLS) and transmission electron microscope (TEM). Optimization of the prepared micelles was conducted based on short-term stability and drug-loading content. The results showed that optimized systems were able to remain stable over 7 days. Compared with Taxotere, DTX-PMs with the same ratio of hydrophilic/hydrophobic chain length displayed similar sustained release behaviors. The cytotoxicity of the optimized targeted DTX-PCL12K-PEG5K-SMLP micelles (DTX-PMs2) and non-targeted DTX-PCL12K-mPEG5K micelles (DTX-PMs1) were evaluated by MTT assays using prostate specific membrane antigen (PSMA) positive prostate adenocarcinoma cells (LNCaP). The results showed that the targeted micelles had a much lower IC50 than their non-targeted counterparts (48 h: 0.87 ± 0.27 vs 13.48 ± 1.03 µg/ml; 72 h: 0.02 ± 0.008 vs 1.35 ± 0.54 µg/ml). In vitro cellular uptake of PMs2 showed 5-fold higher fluorescence intensity than that of PMs1 after 4 h incubation. According to these results, the novel nano-sized drug delivery system based on DTX-PCL-PEG-SMLP offers great promise for the treatment of prostatic cancer.
Sigma-Aldrich
Carboxylic acid-poly(ethylene glycol)-b-poly(ε-caprolactone), PEG average Mn 5000, PCL average Mn 5000
Sigma-Aldrich
Carboxylic acid-poly(ethylene glycol)-b-poly(ε-caprolactone), PEG average Mn 2,000, PCL average Mn 5,000
Sigma-Aldrich
Maleimide-poly(ethylene glycol)-b-poly(ε-caprolactone), PEG average Mn 5000, PCL average Mn 5000