Merck
  • Hepatic lipase affects both HDL and ApoB-containing lipoprotein levels in the mouse.
Hepatic lipase affects both HDL and ApoB-containing lipoprotein levels in the mouse.
Biochimica et biophysica acta (1998-06-19)
S Braschi, N Couture, A Gambarotta, B R Gauthier, C R Coffill, D L Sparks, N Maeda, J R Schultz
ABSTRACT
Transgenic mice were created overproducing a range of human HL (hHL) activities (4-23-fold increase) to further examine the role of hepatic lipase (HL) in lipoprotein metabolism. A 5-fold increase in heparin releasable HL activity was accompanied by moderate (approx. 20%) decreases in plasma total and high density lipoprotein (HDL) cholesterol and phospholipid (PL) but no significant change in triglyceride (TG). A 23-fold increase in HL activity caused a more significant decrease in plasma total and HDL cholesterol, PL and TG (77%, 64%, 60%, and 24% respectively), and a substantial decrease in lipoprotein lipids amongst IDL, LDL and HDL fractions. High levels of HL activity diminished the plasma concentration of apoA-I, A-II and apoE (76%, 48% and 75%, respectively). In contrast, the levels of apoA-IV-containing lipoproteins appear relatively resistant to increased titers of hHL activity. Increased hHL activity was associated with a progressive decrease in the levels and an increase in the density of LpAI and LpB48 particles. The increased rate of disappearance of 125I-labeled human HDL from the plasma of hHL transgenic mice suggests increased clearance of HDL apoproteins in the transgenic mice. The effect of increased HL activity on apoB100-containing lipoproteins was more complex. HL-deficient mice have substantially decreased apoB100-containing low density lipoproteins (LDL) compared to controls. Increased HL activity is associated with a transformation of the lipoprotein density profile from predominantly buoyant (VLDL/IDL) lipoproteins to more dense (LDL) fractions. Increased HL activity from moderate (4-fold) to higher (5-fold) levels decreased the levels of apoB100-containing particles. Thus, at normal to moderately high levels in the mouse, HL promotes the metabolism of both HDL and apoB-containing lipoproteins and thereby acts as a key determinant of plasma levels of both HDL and LDL.
Product Description
G-CAPSULE Accessory Weight
Pricing and availability is not currently available.
G-CAPSULE electroelution device
Pricing and availability is not currently available.