Merck
The ADAMTS13
Haematologica (2017-07-29)
Laurent Gilardin, Sandrine Delignat, Ivan Peyron, Mathieu Ing, Yu-Chun Lone, Bagirath Gangadharan, Baptiste Michard, Yousra Kherabi, Meenu Sharma, Anastas Pashov, Jean-Baptiste Latouche, Mohamad Hamieh, Olivier Toutirais, Pascale Loiseau, Lionel Galicier, Agnès Veyradier, Srini Kaveri, Bernard Maillère, Paul Coppo, Sébastien Lacroix-Desmazes
ABSTRACT
Acquired thrombotic thrombocytopenic purpura is a rare and severe disease characterized by auto-antibodies directed against "A Disintegrin And Metalloproteinase with Thrombospondin type 1 repeats, 13
Sigma-Aldrich
4-Methylumbelliferyl phosphate