- Utility of ultrasound assisted-cloud point extraction and spectophotometry as a preconcentration and determination tool for the sensitive quantification of mercury species in fish samples.
Utility of ultrasound assisted-cloud point extraction and spectophotometry as a preconcentration and determination tool for the sensitive quantification of mercury species in fish samples.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2017-08-16)
Nail Altunay
PMID28810179
ABSTRACT
The current study reports, for the first time, the development of a new analytical method employing ultrasound assisted-cloud point extraction (UA-CPE) for the extraction of CH