Merck
  • Porcine Ischemic Wound-Healing Model for Preclinical Testing of Degradable Biomaterials.
Porcine Ischemic Wound-Healing Model for Preclinical Testing of Degradable Biomaterials.
Tissue engineering. Part C, Methods (2017-08-02)
Prarthana Patil, John R Martin, Samantha M Sarett, Alonda C Pollins, Nancy L Cardwell, Jeffrey M Davidson, Scott A Guelcher, Lillian B Nanney, Craig L Duvall
ABSTRACT
Impaired wound healing that mimics chronic human skin pathologies is difficult to achieve in current animal models, hindering testing and development of new therapeutic biomaterials that promote wound healing. In this article, we describe a refinement and simplification of the porcine ischemic wound model that increases the size and number of experimental sites per animal. By comparing three flap geometries, we adopted a superior configuration (15 × 10 cm) that enabled testing of twenty 1 cm