Merck
  • ER-associated degradation regulates Alzheimer's amyloid pathology and memory function by modulating γ-secretase activity.
ER-associated degradation regulates Alzheimer's amyloid pathology and memory function by modulating γ-secretase activity.
Nature communications (2017-11-15)
Bing Zhu, LuLin Jiang, Timothy Huang, Yingjun Zhao, Tongfei Liu, Yongwang Zhong, Xiaoguang Li, Alexandre Campos, Kenneth Pomeroy, Eliezer Masliah, Dongxian Zhang, Huaxi Xu
ABSTRACT
Endoplasmic-reticulum-associated degradation (ERAD) is an important protein quality control system which maintains protein homeostasis. Constituents of the ERAD complex and its role in neurodegeneration are not yet fully understood. Here, using proteomic and FRET analyses, we demonstrate that the ER protein membralin is an ERAD component, which mediates degradation of ER luminal and membrane substrates. Interestingly, we identify nicastrin, a key component of the γ-secretase complex, as a membralin binding protein and membralin-associated ERAD substrate. We demonstrate a reduction of membralin mRNA and protein levels in Alzheimer's disease (AD) brain, the latter of which inversely correlates with nicastrin abundance. Furthermore, membralin deficiency enhances γ-secretase activity and neuronal degeneration. In a mouse AD model, downregulating membralin results in β-amyloid pathology, neuronal death, and exacerbates synaptic/memory deficits. Our results identify membralin as an ERAD component and demonstrate a critical role for ERAD in AD pathogenesis.
Sigma-Aldrich
Anti-HRD1/SYVN1 antibody produced in rabbit, ~1.0 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Membralin, from rabbit
Sigma-Aldrich
Anti-Nicastrin Antibody, ascites fluid, clone 9C3, Chemicon®
Sigma-Aldrich
Anti-TMEM259 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution