Merck
  • Antitumor activity of the retinoid-related molecules (E)-3-(4'-hydroxy-3'-adamantylbiphenyl-4-yl)acrylic acid (ST1926) and 6-[3-(1-adamantyl)-4-hydroxyphenyl]-2-naphthalene carboxylic acid (CD437) in F9 teratocarcinoma: Role of retinoic acid receptor gamma and retinoid-independent pathways.
Antitumor activity of the retinoid-related molecules (E)-3-(4'-hydroxy-3'-adamantylbiphenyl-4-yl)acrylic acid (ST1926) and 6-[3-(1-adamantyl)-4-hydroxyphenyl]-2-naphthalene carboxylic acid (CD437) in F9 teratocarcinoma: Role of retinoic acid receptor gamma and retinoid-independent pathways.
Molecular pharmacology (2006-06-22)
Edoardo Parrella, Maurizio Giannì, Maddalena Fratelli, Maria Monica Barzago, Ivan Raska, Luisa Diomede, Mami Kurosaki, Claudio Pisano, Paolo Carminati, Lucio Merlini, Sabrina Dallavalle, Michele Tavecchio, Cecile Rochette-Egly, Mineko Terao, Enrico Garattini
ABSTRACT
The retinoid-related molecules (RRMs) ST1926 [(E)-3-(4'-hydroxy-3'-adamantylbiphenyl-4-yl)acrylic acid] and CD437 (6-[3-(1-adamantyl)-4-hydroxyphenyl]-2-naphthalene carboxylic acid) are promising anticancer agents. We compared the retinoic acid receptor (RAR) trans-activating properties of the two RRMs and all-trans-retinoic acid (ATRA). ST1926 and CD437 are better RARgamma agonists than ATRA. We used three teratocarcinoma cell lines to evaluate the significance of RARgamma in the activity of RRMs: F9-wild type (WT); F9gamma-/-, lacking the RARgamma gene; F9gamma51, aF9gamma-/-derivative, complemented for the RARgamma deficit. Similar to ATRA, ST1926 and CD437 activate cytodifferentiation only in F9-WT cells. Unlike ATRA, ST1926 and CD437 arrest cells in the G2/M phase of the cell cycle and induce apoptosis in all F9 cell lines. Our data indicate that RARgamma and the classic retinoid pathway are not relevant for the antiproliferative and apoptotic activities of RRMs in vitro. Increases in cytosolic calcium are fundamental for apoptosis, in that intracellular calcium chelators abrogate the process. Comparison of the gene expression profiles associated with ST1926 and ATRA in F9-WT and F9gamma-/-indicates that the RRM activates a conspicuous nonretinoid response in addition to the classic and RAR-dependent pathway. The pattern of genes regulated by ST1926 selectively, in a RARgamma-independent manner, provides novel insights into the possible molecular determinants underlying the activity of RRMs in vitro. Furthermore, it suggests that RARgamma-dependent responses are relevant to the activity of RRMs in vivo. Indeed, the receptor hinders the antitumor activity in vivo, in that both syngeneic and immunosuppressed SCID mice bearing F9gamma-/- tumors have increased life spans after treatment with ST1926 and CD437 relative to their F9-WT counterparts.
Sigma-Aldrich
ST1926, ≥98% (HPLC)
Pricing and availability is not currently available.