- Exogenous application of poly-γ-glutamic acid enhances stress defense in Brassica napus L. seedlings by inducing cross-talks between Ca
Exogenous application of poly-γ-glutamic acid enhances stress defense in Brassica napus L. seedlings by inducing cross-talks between Ca
Plant physiology and biochemistry : PPB (2017-07-26)
Zongqi Xu, Peng Lei, Xiao Pang, Huashan Li, Xiaohai Feng, Hong Xu
PMID28743039
ABSTRACT
Poly-γ-glutamic acid (γ-PGA) is a microbe-secreted isopeptide shown to promote growth and enhance crop stress tolerance. However, its downstream signaling pathways are unknown. Here, we studied γ-PGA-induced tolerance to salt and cold stresses. Pretreatment with γ-PGA contributed to enhance stress tolerance of canola seedlings by promoting proline accumulation and total antioxidant capacity (T-AOC) improvement. Further, Ca