Merck
  • Synergistic Activity with NOTCH Inhibition and Androgen Ablation in ERG-Positive Prostate Cancer Cells.
Synergistic Activity with NOTCH Inhibition and Androgen Ablation in ERG-Positive Prostate Cancer Cells.
Molecular cancer research : MCR (2017-06-14)
Ahmed A Mohamed, Shyh-Han Tan, Charles P Xavier, Shilpa Katta, Wei Huang, Lakshmi Ravindranath, Muhammad Jamal, Hua Li, Meera Srivastava, Eri S Srivatsan, Taduru L Sreenath, David G McLeod, Alagarsamy Srinivasan, Gyorgy Petrovics, Albert Dobi, Shiv Srivastava
ABSTRACT
The oncogenic activation of the ETS-related gene (
Sigma-Aldrich
MISSION® esiRNA, targeting human ERG
Sigma-Aldrich
DAPT, ≥98% (HPLC), solid