Merck
  • A solvent- and vacuum-free route to large-area perovskite films for efficient solar modules.
A solvent- and vacuum-free route to large-area perovskite films for efficient solar modules.
Nature (2017-09-05)
Han Chen, Fei Ye, Wentao Tang, Jinjin He, Maoshu Yin, Yanbo Wang, Fengxian Xie, Enbing Bi, Xudong Yang, Michael Grätzel, Liyuan Han
ABSTRACT
Recent advances in the use of organic-inorganic hybrid perovskites for optoelectronics have been rapid, with reported power conversion efficiencies of up to 22 per cent for perovskite solar cells. Improvements in stability have also enabled testing over a timescale of thousands of hours. However, large-scale deployment of such cells will also require the ability to produce large-area, uniformly high-quality perovskite films. A key challenge is to overcome the substantial reduction in power conversion efficiency when a small device is scaled up: a reduction from over 20 per cent to about 10 per cent is found when a common aperture area of about 0.1 square centimetres is increased to more than 25 square centimetres. Here we report a new deposition route for methyl ammonium lead halide perovskite films that does not rely on use of a common solvent or vacuum: rather, it relies on the rapid conversion of amine complex precursors to perovskite films, followed by a pressure application step. The deposited perovskite films were free of pin-holes and highly uniform. Importantly, the new deposition approach can be performed in air at low temperatures, facilitating fabrication of large-area perovskite devices. We reached a certified power conversion efficiency of 12.1 per cent with an aperture area of 36.1 square centimetres for a mesoporous TiO
Sigma-Aldrich
Methylamine solution, 40 wt. % in H2O
Pricing and availability is not currently available.