Merck
  • Up-regulation of gap junction in peripheral blood T lymphocytes contributes to the inflammatory response in essential hypertension.
Up-regulation of gap junction in peripheral blood T lymphocytes contributes to the inflammatory response in essential hypertension.
PloS one (2017-09-15)
Xin Ni, Ai Wang, Liang Zhang, Li-Ya Shan, Hai-Chao Zhang, Li Li, Jun-Qiang Si, Jian Luo, Xin-Zhi Li, Ke-Tao Ma
ABSTRACT
Inflammation has been shown to play an important role in the mechanisms involved in the pathogenesis of hypertension. Connexins (Cxs)-based gap junction channels (GJCs) or hemichannels (HCs) are involved in the maintenance of homeostasis in the immune system. However, the role of Cx43-based channels in T-lymphocytes in mediating the immune response in essential hypertension is not fully understand. The present study was designed to investigate the role of Cxs-based channels in T lymphocytes in the regulation of hypertension-mediated inflammation. The surface expressions of T lymphocyte subtypes, Cx40/Cx43, and inflammatory cytokines (IFN-γ (interferon-gamma) and TNF-ɑ (tumor necrosis factor alpha)) in T cells, as well as gap junction communication of peripheral blood lymphocytes from essential hypertensive patients (EHs) and normotensive healthy subjects (NTs) were detected by flow cytometry. Expression levels and phosphorylation of Cx43 protein in peripheral blood lymphocytes of EHs and NTs were analyzed by Western blot. The proliferation rate of peripheral blood mononuclear cells (PBMCs) after treatment with a Cxs inhibitor was examined by a CCK-8 assay. The levels of inflammatory cytokines were detected using ELISA. Within the CD3+ T cell subsets, we found a significant trend toward an increase in the percentage of CD4+ T cells and CD4+/CD8+ ratio as well as in serum levels of IFN-γ and TNF-ɑ in the peripheral blood of EHs compared with those in NTs. Moreover, the peripheral blood lymphocytes of EH patients exhibited enhanced GJCs formation, increased Cx43 protein level and Cx43 phosphorylation at Ser368, and a significant increase in Cx40/Cx43 surface expressions levels in CD4+ or CD8+ T lymphocytes. Cx43-based channel inhibition by a mimetic peptide greatly reduced the exchange of dye between lymphocytes, proliferation of stimulated lymphocytes and the pro-inflammatory cytokine levels of EHs and NTs. Our data suggest that Cx40/Cx43-based channels in lymphocytes may be involved in the regulation of T lymphocyte proliferation and the production of pro-inflammatory cytokines, which contribute to the hypertensive inflammatory response.
Product Description
Ficoll®-Paque Premium, Cytiva 17-5442-02, pack of 6 × 100 mL