Merck
  • Manganese-induced trafficking and turnover of GPP130 is mediated by sortilin.
Manganese-induced trafficking and turnover of GPP130 is mediated by sortilin.
Molecular biology of the cell (2017-08-05)
Swati Venkat, Adam D Linstedt
ABSTRACT
Elevated, nontoxic doses of manganese (Mn) protect against Shiga toxin-1-induced cell death via down-regulation of GPP130, a cycling Golgi membrane protein that serves as an endosome-to-Golgi trafficking receptor for the toxin. Mn binds to GPP130 in the Golgi and causes GPP130 to oligomerize/aggregate, and the complexes are diverted to lysosomes. In fact, based on experiments using the self-interacting FM domain, it appears generally true that aggregation of a Golgi protein leads to its lysosomal degradation. How such oligomers are selectively sorted out of the Golgi is unknown. Here we provide evidence that Mn-induced exit of GPP130 from the
Sigma-Aldrich
Cycloheximide, from microbial, ≥94% (TLC)
Sigma-Aldrich
Anti-γ-Tubulin antibody, Mouse monoclonal, clone GTU-88, ascites fluid
Sigma-Aldrich
Anti-HA antibody, Mouse monoclonal, clone HA-7, purified from hybridoma cell culture
Sigma-Aldrich
Monoclonal Anti-GFP antibody produced in mouse, clone GT859, affinity isolated antibody
Sigma-Aldrich
MISSION® esiRNA, targeting human SORT1
Sigma-Aldrich
Neurotensin, ≥90% (HPLC)