Merck
  • Downregulation of type 3 inositol (1,4,5)-trisphosphate receptor decreases breast cancer cell migration through an oscillatory Ca
Downregulation of type 3 inositol (1,4,5)-trisphosphate receptor decreases breast cancer cell migration through an oscillatory Ca
Oncotarget (2017-10-27)
Abdallah Mound, Alexia Vautrin-Glabik, Arthur Foulon, Béatrice Botia, Frédéric Hague, Jan B Parys, Halima Ouadid-Ahidouch, Lise Rodat-Despoix
ABSTRACT
Breast cancer remains a research priority due to its invasive phenotype. Although the role of ion channels in cancer is now well established, the role of inositol (1,4,5)-trisphosphate (IP
Sigma-Aldrich
MISSION® esiRNA, targeting human ITPR3
Pricing and availability is not currently available.
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) assay mix, lyophilized powder
Pricing and availability is not currently available.
Sigma-Aldrich
Adenosine 5′-triphosphate (ATP) bioluminescent somatic cell assay kit, for cellular ATP determination
Pricing and availability is not currently available.
Sigma-Aldrich
Somatic cell ATP releasing reagent, for ATP determination
Pricing and availability is not currently available.