Merck
  • Anticancer effect of aloe-emodin on cervical cancer cells involves G2/M arrest and induction of differentiation.
Anticancer effect of aloe-emodin on cervical cancer cells involves G2/M arrest and induction of differentiation.
Acta pharmacologica Sinica (2007-11-23)
Jun-ming Guo, Bing-xiu Xiao, Qiong Liu, Shun Zhang, Dong-hai Liu, Zhao-hui Gong
ABSTRACT
The aim of this study was to investigate the effects of aloe-emodin, a natural compound from the root and rhizome of Rheum palmatum, on the growth of human cervical cancer cells, HeLa. HeLa cells were treated with various concentrations of aloe-emodin for 1-5 d, and cell growth was measured by 3-(4, 5-dimethylthiazol-2-yl)-2, 5-diphenyl tetrazolium bromide assay. The long-term growth effect was investigated by crystal violet assay. The distributions of the cell cycle and apoptosis were analyzed by flow cytometry. The alkaline phosphatase (ALP) activity was analyzed by a chemical analyzer. Finally, Western blotting was used to indicate the abundant changes of protein kinase C (PKC), c-myc, cyclins, cyclin-dependent kinases (CDK), and proliferating cell nuclear antigen (PCNA). Aloe-emodin inhibited the growth of HeLa cells in a dose-dependent manner at concentrations ranging between 2.5 and 40 micromol/L. The flow cytometric analysis showed that HeLa cells were arrested at the G2/M phase. This effect was associated with the decrease in cyclin A and CDK2, and the increase in cyclin B1 and CDK1. More importantly, the ALP activity was found to be increased by aloe-emodin treatment, and accompanied by the inhibition of PCNA expression. In addition, aloe-emodin suppressed the expression of PKCalpha and c-myc. These findings provide a possible mechanistic explanation for the growth inhibitory effect of aloe-emodin on HeLa, which includes cell cycle arrest and inducing differentiation.
Sigma-Aldrich
Aloe-emodin, ≥95% (HPLC)
Pricing and availability is not currently available.