Merck
  • The ecto-ATPase inhibitor ARL 67156 enhances parasympathetic neurotransmission in the guinea-pig urinary bladder.
The ecto-ATPase inhibitor ARL 67156 enhances parasympathetic neurotransmission in the guinea-pig urinary bladder.
European journal of pharmacology (1997-06-25)
T D Westfall, C Kennedy, P Sneddon
ABSTRACT
The influence of enzymatic degradation on the neurotransmitter actions of ATP was studied using the ecto-ATPase inhibitor 6-N,N-diethyl-D-beta,gamma-dibromomethyleneATP (ARL 67156). Field stimulation of the parasympathetic nerves innervating guinea-pig urinary bladder muscle strips (1-8 Hz for 20 s) produced characteristic biphasic contractions, the peak magnitudes of which were significantly increased by 29-32% by ARL 67156 (100 microM). A similar degree of enhancement was seen in the presence of atropine (1 microM), consistent with ARL 67156 acting to enhance the action of neuronally released ATP. The effects of ARL 67156 reversed rapidly on washout of the drug. Contractions evoked by exogenous ATP (100 microM) were also potentiated by ARL 67156 (100 microM), but those to the stable analogue alpha,beta-methyleneATP (5 microM) were unaffected. ARL 67156 (100 microM) also enhanced contractions to exogenous acetylcholine (1 microM) and histamine (3 microM), but this potentiation was abolished by pyridoxalphosphate-6-azophenyl-2',4'-disulphonic acid (PPADS) (100 mciroM). It is concluded that when ATP acts as a neurotransmitter its postjunctional actions are attenuated by enzymatic degradation. ARL 67156 inhibits this breakdown.
Sigma-Aldrich
ARL 67156 trisodium salt hydrate, ≥98% (HPLC), solid
Pricing and availability is not currently available.