Merck
  • Interfacially Engineered Pyridinium Pseudogemini Surfactants as Versatile and Efficient Supramolecular Delivery Systems for DNA, siRNA, and mRNA.
Interfacially Engineered Pyridinium Pseudogemini Surfactants as Versatile and Efficient Supramolecular Delivery Systems for DNA, siRNA, and mRNA.
ACS applied materials & interfaces (2017-08-16)
Uttam Satyal, Bogdan Draghici, Lisa L Dragic, Qiangnan Zhang, Kyle W Norris, Muniswamy Madesh, Eugen Brailoiu, Marc A Ilies
ABSTRACT
This article presents the synthesis, self-assembly, and biological activity as transfection agents for pDNA, siRNA, and mRNA of novel pyridinium pseudogemini surfactants, interfacially engineered from the most efficient gemini surfactants and lipids generated in our amphiphile research program. Formulation of novel amphiphiles in water revealed supramolecular properties very similar to those of gemini surfactants, despite their lipidlike charge/mass ratio. This dual character was found also to enhance endosomal escape and significantly increase the transfection efficiency. We were also successful in identifying the parameters governing the efficient delivery of pDNA, siRNA, and mRNA, drawing valuable structure-activity and structure-property relationships for each nucleic acid type, and establishing DNA/siRNA/mRNA comparisons. Several supramolecular complexes identified in this study proved to be extremely efficient nucleic acid delivery systems, displaying excellent serum stability and tissue penetration in three-dimensional organoids.
Sigma-Aldrich
MISSION® esiRNA, targeting human PLG
Sigma-Aldrich
Tetradecylamine, 95%
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder